Pyramex™ V2G™ Safety Eyewear
Foam padding forms a perfect seal to keep out dust and airborne particles.
Supplier: Pyramex™ GB1820ST
Description
- Lenses are treated to prevent fogging and self seals small scratches
- 9.75 base curved lens provides full side protection
- Headband features quick fit retainer
- Lenses provide 99% protection against harmful UV rays
Specifications
Scratch-resistant | |
Safety goggles | |
Polycarbonate |
Gray | |
Black strap; Foam padding; Interchangeable temples | |
0.999 |