missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mettler Toledo™ V10S Compact Volumetric KF Titrator

Ideal for fast and precise water content determination with easy operation. Mettler Toledo V10S Compact Volumetric KF Titrators can analyze samples with water content between 100 ppm and 100% in single measurements or sample series with statistics.

Supplier:  Mettler Toledo™ 30267116

Catalog No. 01-914-315

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

One Click™ User Interface
  • Large color touchscreen (VGA 5.7 in. color TFT, 640 x 480 pixel) with simple-to-understand and fast-to-operate One Click user interface
  • All users have their own homescreen with dedicated shortcut buttons (4 per user) to directly start specific routine tasks
  • User interface in English, German, French, Spanish, Italian, Chinese, Russian, Polish, Korean and Portuguese available
Solvent Manager for Safe Liquid Handling
  • Protects users against contact with chemicals
  • Takes care of filling, draining and exchange of reagents
  • It is controlled by the titrator and offers solvent exchange with One Click without additional user interaction
Intelligent Burettes
  • Automatically recognizes titrant when the burette is installed on the titrator
  • Important data is saved on the RFID chip and directly transferred to the titrator, where it is stored and can be monitored
  • Eliminates usage of wrong or an expired titrant
Flexible Data Export
  • Broad range of options for handling data
  • Option to export as CSV, PDF or XML files
  • Printers, USB barcode reader or USB memory sticks can be connected via the USB port and are automatically recognized
  • Common USB printers ( DIN-A4/US letter) or the USB-P25 compact printer for secure and GLP-compliant paper recording
Communication
  • Data Export: USB, Ethernet, PDF
  • Ethernet (TCP/IP): 1
  • COM (serial balance connection): 1
  • USB host (printer, memory stick, barcode reader, hub): 2
  • Integrated Magnetic Stirrer: 1
  • Pump/Stirrer: 1
Methods and Series
  • KF standard methods: 2
  • Max. number of user methods: 5
  • Number of method functions: 10
  • Number of samples per series: 120
  • Autostart: automatic start after sample addition: Yes
  • Drift stop for endpoint recognition (absolute and relative, tmax, delay): yes
  • Max. number of calculations/results per method: 3
  • Statistics of up to 120 samples including result limits check, outlier test: Yes
  • Recalculation of results: Yes
  • Data in Online Screen: time, water count, potential, stirrer speed, volume, drift: Yes
  • Data in measured value table: time, water count, potential, volume, drift: Yes
  • Curves in Online Screen and printout: E-V, E-t, V-t, drift-t, water count-t: Yes
Specifications
  • Measurement Range: -2000 to +2000mV
  • Resolution/Limits of Error: 0.1mV/2mV
  • Polarization Current Range: 0 to 24μA
  • Polarization Current Resolution/Limits of Error: 0.1μA/1μA
  • Burette Drive Resolution: 20,000 steps
  • Burette Resolution (for 5mL burette) 0.25μL
  • Burette Limits of Error: 0.3% of the respective volume
  • Burette Resolver: 32 pulses/mm stroke
  • Burette Resolver Resolution: 0.0625% of burette volume
  • Filling Time and Discharge Time: 20 sec. at 100% filling rate
  • Ambient Temperature: 5° to 40°C
  • Atmospheric Humidity: max. 80% at 31°C
  • Drift Measurement: Online, <5μg/min.
  • Measurement Range: 100 ppm to 100%
  • Repeatability: 0.3% at >10mg water
  • Glass Beaker: 150mL
  • Dimensions (D x W x H): 33.3 x 21 x 32cm
  • Weight: 4.2kg

Certifications

CE, ISO 9001, ISO 14001

Specifications

Specifications

CE, ISO 9001, ISO 14001
Online, <5 μg/min.
V10S
0.1mV/2mV; 0.1μA/1μA; 0.1μg
5°C to 40°C
21 cm
0 to 24μA (Polarization Current Range)
VGA 5.7 in. color TFT, 640 x 480 pixel
-2000 to +2000 mV
120 (per series)
Volumetric KF Titrator
33.3 cm
SDS
Documents

Documents

Promotions

Promotions