Please login to your online account to display your discounted pricing

V gamma 2 TCR Hamster anti-Mouse, PE-Cy7, Clone: UC3-10A6, eBioscience™

Hamster Monoclonal Antibody

$158.05 - $361.05

Specifications

Antigen V gamma 2 TCR
Clone UC3-10A6
Host Species Hamster
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123391

 
life technologies
LS25582880
25μg Each for $158.05

501123392

 
life technologies
LS25582882
100μg Each for $361.05
Description & Specifications

Specifications

Antigen V gamma 2 TCR
Clone UC3-10A6
Host Species Hamster
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The UC3-10A6 monoclonal antibody reacts with the mouse V gamma 2 (Vg2) delta chain of the gamma delta T cell receptor (TCR). The gamma delta TCR is expressed by a small subset of T cells in the thymus, peripheral lymphoid tissues, intestinal epithelium, and epidermis. The exact specificity, ligand and function of gamma delta TCR-bearing T cells are not yet fully understood. Studies suggest that these cells recognize bacterial ligands and some tumor cells in the context of MHC class I-like gene products and play a role in regulating the immune response during bacterial infection.