Please login to your online account to display your discounted pricing

V gamma 2 TCR Armenian Hamster anti-Mouse, PerCP-eFluor 710, Clone: UC3-10A6, eBioscience™

Armenian Hamster Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen V gamma 2 TCR
Clone UC3-10A6
Host Species Armenian Hamster
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123838

 
affymetrix
46582880
25μg Each for $172.55

501129077

 
affymetrix
46582882
100μg Each for $433.55
Description & Specifications

Specifications

Antigen V gamma 2 TCR
Clone UC3-10A6
Host Species Armenian Hamster
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The UC3-10A6 monoclonal antibody reacts with the mouse V gamma 2 (Vg2) delta chain of the gamma delta T cell receptor (TCR). The gamma delta TCR is expressed by a small subset of T cells in the thymus, peripheral lymphoid tissues, intestinal epithelium, and epidermis. The exact specificity, ligand and function of gamma delta TCR-bearing T cells are not yet fully understood. Studies suggest that these cells recognize bacterial ligands and some tumor cells in the context of MHC class I-like gene products and play a role in regulating the immune response during bacterial infection.