Please login to your online account to display your discounted pricing

V Beta 9 TCR Mouse anti-Human, APC, Clone: AMKB1-2, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $375.55

Specifications

Antigen V Beta 9 TCR
Clone AMKB1-2
Host Species Mouse
Gene Alias Vbeta 9, Vb9
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015181

 
life technologies
LS17489941
25 tests Each for $172.55

5015182

 
life technologies
LS17489942
100 tests Each for $375.55
Description & Specifications

Specifications

Antigen V Beta 9 TCR
Clone AMKB1-2
Host Species Mouse
Gene Alias Vbeta 9, Vb9
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

This AMKB1-2 monoclonal antibody recognizes the human T cell receptor (TCR) Vβ9 allele. Composed of an α and β chain, TCR specificity is typically determined by Vα, Jα, Vβ, Dβ, and Jβ gene rearrangement. Vβ expression in humans has been examined in studies on the effects of superantigens, inflammation, autoimmune disease, and HIV infection. More recently, assessment of TCR Vβ expression has been used to phenotype T cell clonality in CD3+/TCRαβ+ large granular lymphocyte leukemias. A member of the Ig superfamily, this receptor is expressed on a subset of peripheral blood T cells.