Please login to your online account to display your discounted pricing

V beta 8.1, 8.2 TCR Rat anti-Mouse, PerCP-eFluor 710, Clone: KJ-16, eBioscience™

Rat Monoclonal Antibody

$158.05 - $477.05

Specifications

Antigen V beta 8.1, 8.2 TCR
Clone KJ-I6)
Host Species Rat
Gene Alias Vbeta8.1, Vb 8.1, Vb8.2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016083

 
affymetrix
46581380
25μg Each for $158.05

5016084

 
affymetrix
46581382
100μg Each for $477.05
Description & Specifications

Specifications

Antigen V beta 8.1, 8.2 TCR
Clone KJ-I6)
Host Species Rat
Gene Alias Vbeta8.1, Vb 8.1, Vb8.2
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This KJ16-133 monoclonal antibody recognizes mouse T cell receptors (TCR) containing Vβ 8.1 or 8.2, which is expressed by 15-20% of peripheral T cells in most strains of mice (ie. Balb/c, AKR, C57BL/6, B10.A) and by a number of T cell hybridomas (ie. DO-11.10, 3DT-52.5, D1G10G11, SKK-9.11, SKK-45.10, MDK16, S18.4).