Please login to your online account to display your discounted pricing

V beta 6 TCR Rat anti-Mouse, APC, Clone: RR4-7, eBioscience™

Rat Monoclonal Antibody

$143.55 - $332.05

Specifications

Antigen V beta 6 TCR
Clone RR4-7
Host Species Rat
Gene Alias Vb6-TCR, Vb-6
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245690

 
life technologies
LS17579580
25μg Each for $143.55

50245691

 
life technologies
LS17579582
100μg Each for $332.05
Description & Specifications

Specifications

Antigen V beta 6 TCR
Clone RR4-7
Host Species Rat
Gene Alias Vb6-TCR, Vb-6
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This RR4-7 monoclonal antibody reacts with mouse V beta 6 TCR. Composed of an a and b chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The RR4-7 antibody recognizes the V beta 6 chain on T cells from mouse strains with the a (e.g., C57BR and SJL) and b haplotypes (e.g., BALB/c, C3H, and DBA/1). Reports indicate that strains expressing the Mls-1a antigen, particularly in the presence of MHC Class II I-E, possess fewer V beta 6 TCR+ T cells.

The RR4-7 antibody has been reported to have functional activity in vitro and in vivo.