missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

V beta 5.1 TCR Mouse anti-Human, PE-Cy7, Clone: LC4, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $421.08

Specifications

Antigen V beta 5.1 TCR
Clone LC4
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123395 Life Technologies
LS25583241
25 Tests Each for $172.55
501123396 Life Technologies
LS25583242
100 Tests Each for $421.08
Specifications

Description

This LC4 monoclonal antibody recognizes the human T cell receptor (TCR) Vβ5.1 allele. Composed of an α and β chain, TCR specificity is typically determined by Vα, Jα, Vβ, Dβ, and Jβ gene rearrangement. Vβ expression in humans has been examined in studies on the effects of superantigens, inflammation, autoimmune disease, and HIV infection. More recently, assessment of TCR Vβ expression has been used to phenotype T cell clonality in CD3+/TCRαβ+ large granular lymphocyte leukemias. A member of the Ig superfamily, this receptor is expressed on a subset of peripheral blood T cells.

This monoclonal antibody has been reported to induce apoptosis and calcium flux.
Description & Specifications

Specifications

V beta 5.1 TCR
Murine
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.25μg)/test
LC4
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer