Please login to your online account to display your discounted pricing

V beta 5.1 TCR Mouse anti-Human, APC, Clone: LC4, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen V beta 5.1 TCR
Clone LC4
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015221

 
life technologies
LS17583241
25 Tests Each for $172.55

5015222

 
life technologies
LS17583242
100 Tests Each for $433.55
Description & Specifications

Specifications

Antigen V beta 5.1 TCR
Clone LC4
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

This LC4 monoclonal antibody recognizes the human T cell receptor (TCR) Vβ5.1 allele. Composed of an α and β chain, TCR specificity is typically determined by Vα, Jα, Vβ, Dβ, and Jβ gene rearrangement. Vβ expression in humans has been examined in studies on the effects of superantigens, inflammation, autoimmune disease, and HIV infection. More recently, assessment of TCR Vβ expression has been used to phenotype T cell clonality in CD3+/TCRαβ+ large granular lymphocyte leukemias. A member of the Ig superfamily, this receptor is expressed on a subset of peripheral blood T cells.

This monoclonal antibody has been reported to induce apoptosis and calcium flux.