Please login to your online account to display your discounted pricing

V beta 5.1/5.2 TCR Mouse anti-Mouse, PerCP-eFluor 710, Clone: MR9-4, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $506.05

Specifications

Antigen V beta 5.1/5.2 TCR
Clone MR9-4
Host Species Murine
Gene Alias Vb5.1, Vb5.2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245913

 
life technologies
LS46579680
25μg Each for $201.55

50245914

 
life technologies
LS46579682
100μg Each for $506.05
Description & Specifications

Specifications

Antigen V beta 5.1/5.2 TCR
Clone MR9-4
Host Species Murine
Gene Alias Vb5.1, Vb5.2
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This MR9-4 monoclonal antibody reacts with the mouse T cell receptor (TCR) V beta 5.1/5.2 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The MR9-4 antibody recognizes the V beta 5.1 and 5.2 chains on T cells from mouse strains with the b haplotype of the Tcrb gene, including C57BL/6, B10, and C58/J. V beta 5.1 and 5.2 are deleted in mouse strains that express MHC Class II I-E, including Balb/c, C3H, and DBA/2. Reports also indicate that V beta 5+ T cells recognize the OVA peptide.