missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

V beta 5.1/5.2 TCR Mouse anti-Mouse, eFluor 450, Clone: MR9-4, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $404.55

Specifications

Antigen V beta 5.1/5.2 TCR
Clone MR9-4
Host Species Murine
Gene Alias Vb5.1, Vb5.2
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124174 Life Technologies
LS48579680
25μg Each for $172.55
501129080 Life Technologies
LS48579682
100μg Each for $404.55
Specifications

Description

This MR9-4 monoclonal antibody reacts with the mouse T cell receptor (TCR) V beta 5.1/5.2 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The MR9-4 antibody recognizes the V beta 5.1 and 5.2 chains on T cells from mouse strains with the b haplotype of the Tcrb gene, including C57BL/6, B10, and C58/J. V beta 5.1 and 5.2 are deleted in mouse strains that express MHC Class II I-E, including Balb/c, C3H, and DBA/2. Reports also indicate that V beta 5+ T cells recognize the OVA peptide.
Description & Specifications

Specifications

V beta 5.1/5.2 TCR
Murine
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MR9-4
Vb5.1, Vb5.2
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL