missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

V beta 2 TCR Rat anti-Mouse, FITC, Clone: B20.6, eBioscience™

Rat Monoclonal Antibody

$143.55 - $346.55

Specifications

Antigen V beta 2 TCR
Clone B20.6
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245242 Life Technologies
LS11580480
25μg Each for $143.55
50245243 Life Technologies
LS11580482
100μg Each for $346.55
Specifications

Description

The B20.6 monoclonal antibody reacts with the mouse T-cell receptor (TCR) V beta 2 chain. Neonatal T cells which express V beta 2, and are exposed to the Balb/cV virus are eliminated during negative selection, while adult T cells expressing V beta 2 and exposed to the Balb/cV virus are clonally expanded.
Description & Specifications

Specifications

V beta 2 TCR
Rat
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL
B20.6
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer