missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

V beta 13 TCR Mouse anti-Mouse, APC, Clone: MR12-3, eBioscience™

Mouse Monoclonal Antibody

$143.88 - $368.28

Specifications

Antigen V beta 13 TCR
Clone MR12-3
Host Species Murine
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123141 Life Technologies
LS17579780
25μg Each for $143.88
501123142 Life Technologies
LS17579782
100μg Each for $368.28
Specifications

Description

This MR12-3 monoclonal antibody reacts with the T cell receptor (TCR) V beta 13 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The MR12-3 antibody recognizes the V beta 13 chains on T cells from mouse strains with the b haplotype of the Tcrb gene, including C57BL/6, B10, and C3H. V beta 13+ T cells are deleted in mouse strains with the a (e.g., SJL) and c (e.g., RIIIS/J) haplotypes.

The MR12-3 monoclonal antibody has been reported to have blocking activity.
Description & Specifications

Specifications

V beta 13 TCR
Murine
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
MR12-3
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer