Please login to your online account to display your discounted pricing

V beta 13.2 TCR Mouse anti-Human, PE, Clone: H132, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $361.05

Specifications

Antigen V beta 13.2 TCR
Clone H132
Host Species Murine
Gene Alias Vbeta 13.2, Vbeta13.2, Vb13.2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010872

 
life technologies
LS12580241
25 Tests Each for $158.05

5010873

 
life technologies
LS12580242
100 Tests Each for $361.05
Description & Specifications

Specifications

Antigen V beta 13.2 TCR
Clone H132
Host Species Murine
Gene Alias Vbeta 13.2, Vbeta13.2, Vb13.2
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This H132 monoclonal antibody recognizes the human T cell receptor (TCR) Vβ13.2 allele. Composed of an α and β chain, TCR specificity is typically determined by Vα, Jα, Vβ, Dβ, and Jβ gene rearrangement. Vβ expression in humans has been examined in studies on the effects of superantigens, inflammation, autoimmune disease, and HIV infection. More recently, assessment of TCR Vβ expression has been used to phenotype T cell clonality in CD3+/TCRαβ+ large granular lymphocyte leukemias. A member of the Ig superfamily, this receptor is expressed on a subset of peripheral blood T cells.

The H132 monoclonal antibody recognizes TCR Vβ13.2 specifically, but not Vβ13.1 or Vβ13.3.