Please login to your online account to display your discounted pricing

V beta 10b TCR Rat anti-Mouse, Biotin, Clone: B21.5, eBioscience™

Rat Monoclonal Antibody

$129.05 - $230.55

Specifications

Antigen V beta 10b TCR
Clone B21.5
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011927

 
life technologies
LS13580580
25μg Each for $129.05

5011928

 
life technologies
LS13580582
100μg Each for $230.55
Description & Specifications

Specifications

Antigen V beta 10b TCR
Clone B21.5
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The B21.5 monoclonal antibody reacts with the Vβ10 T Cell Receptor (TCR) in mice of the b haplotype of the TCR Vβ gene complex, which includes AKR, BALB/c, CBA, C3H/He, C57BL, DBA/1 and DBA/2 mice. It does not react with Vβ10 in mice of the a or c haplotypes of the TCR Vβ gene complex, including C57BR, C57L, SJL, and SWR mice.