missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

V alpha 24 J alpha 18 TCR Mouse anti-Human, PerCP-eFluor 710, Clone: 6B11, eBioscience™

Mouse Monoclonal Antibody

$223.08 - $473.88

Specifications

Antigen V alpha 24 J alpha 18 TCR
Clone 6B11
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016079 Life Technologies
LS46580641
25 Tests Each for $223.08
5016080 Life Technologies
LS46580642
100 Tests Each for $473.88
Specifications

Description

The 6B11 monoclonal antibody reacts with an epitope of the CDR3 formed by the germ-line configuration of the V alpha 24 and J alpha 18 of the TCR alpha locus, which results in the expression of an invariant T-cell receptor (TCR) expressed by invariant NK T cells (iNKT). This TCR is homologous to the murine V alpha 14 J alpha 18. This TCR alpha chain pairs with a restricted set of rearranged TCR beta chains, with V beta 11 being the most prominent in humans. NK T cells are restricted by the CD1d molecule and are activated by a CD1d-presented glycolipid ligand, which results in the rapid production of IL-4, IL-13 and IFN gamma. Although iNKT cells have NK-like cytolytic activity, they are considered regulators of immune responses because they rapidly produce large amounts of both Th1 and Th2 cytokines in autoimmune disease, infectious disease, and cancer. The number of iNKT cells present in peripheral blood of normal humans shows considerable variability ranging from 0.01-1% of total peripheral T cells.
Description & Specifications

Specifications

V alpha 24 J alpha 18 TCR
Murine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
6B11
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test