missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

V alpha 2 TCR Rat anti-Mouse, PE-Cy7, Clone: B20.1, eBioscience™

Rat Monoclonal Antibody

$170.28 - $415.80

Specifications

Antigen V alpha 2 TCR
Clone B20.1
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123390 Life Technologies
LS25581282
100μg Each for $415.80
501129118 Life Technologies
LS25581280
25μg Each for $170.28
Specifications

Description

The B20.1 monoclonal antibody reacts with mouse T-cell receptor (TCR) V alpha 2 chains. This antibody recognizes the majority of the TCR V alpha 2 subfamily in mice carrying the a, b and c haplotypes. The antibody also reacts with the products of T cell receptor, V delta 8 due to the high degree of homology.
Description & Specifications

Specifications

V alpha 2 TCR
Rat
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL
B20.1
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer