Please login to your online account to display your discounted pricing

V alpha 2 TCR Rat anti-Mouse, PE, Clone: B20.1, eBioscience™

Rat Monoclonal Antibody

$114.55 - $259.55

Specifications

Antigen V alpha 2 TCR
Clone B20.1
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010888

 
life technologies
LS12581280
25μg Each for $114.55

5010889

 
life technologies
LS12581282
100μg Each for $259.55
Description & Specifications

Specifications

Antigen V alpha 2 TCR
Clone B20.1
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The B20.1 monoclonal antibody reacts with mouse T-cell receptor (TCR) V alpha 2 chains. This antibody recognizes the majority of the TCR V alpha 2 subfamily in mice carrying the a, b and c haplotypes. The antibody also reacts with the products of T cell receptor, V delta 8 due to the high degree of homology.