missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

V alpha 2 TCR Rat anti-Mouse, eFluor 450, Clone: B20.1, eBioscience™

Rat Monoclonal Antibody

$132.00 - $335.28

Specifications

Antigen V alpha 2 TCR
Clone B20.1
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50246013 Life Technologies
LS48581280
25μg Each for $132.00
50246014 Life Technologies
LS48581282
100μg Each for $335.28
Specifications

Description

The B20.1 monoclonal antibody reacts with mouse T-cell receptor (TCR) V alpha 2 chains. This antibody recognizes the majority of the TCR V alpha 2 subfamily in mice carrying the a, b and c haplotypes. The antibody also reacts with the products of T cell receptor, V delta 8 due to the high degree of homology.
Description & Specifications

Specifications

V alpha 2 TCR
Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
B20.1
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL