Please login to your online account to display your discounted pricing

V alpha 11 TCR Rat anti-Mouse, PerCP-eFluor 710, Clone: RR8-1, eBioscience™

Rat Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen V alpha 11 TCR
Clone RR8-1
Host Species Rat
Gene Alias Valpha11, Va11
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123835

 
life technologies
LS46580080
25μg Each for $172.55

501123836

 
life technologies
LS46580082
100μg Each for $433.55
Description & Specifications

Specifications

Antigen V alpha 11 TCR
Clone RR8-1
Host Species Rat
Gene Alias Valpha11, Va11
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This RR8-1 monoclonal antibody reacts with the mouse T cell receptor (TCR) V alpha 11 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The RR8-1 antibody recognizes the V beta 11 chain on T cells from mouse strains with the b (e.g., C57BL/6, B10) and d (e.g., DBA/1, DBA/2) haplotypes. V alpha 11+ T cells are absent in strains with the a (e.g., BALB/c, C3H) or c (e.g., SJL) haplotypes of the Tcra gene complex. Finally, V alpha 11 is expressed predominantly on CD4+ T cells.

The RR8-1 antibody recognizes the V alpha 11.1 and 11.2 alleles, but not 11.3. This antibody has been reported to stimulate V alpha 11+ T cells.