missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

V alpha 11 TCR Rat anti-Mouse, APC, Clone: RR8-1, eBioscience™

Rat Monoclonal Antibody

$143.88 - $328.68

Specifications

Antigen V alpha 11 TCR
Clone RR8-1
Host Species Rat
Gene Alias Valpha11, Va11
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245694 Life Technologies
LS17580080
25μg Each for $143.88
50245695 Life Technologies
LS17580082
100μg Each for $328.68
Specifications

Description

This RR8-1 monoclonal antibody reacts with the mouse T cell receptor (TCR) V alpha 11 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The RR8-1 antibody recognizes the V beta 11 chain on T cells from mouse strains with the b (e.g., C57BL/6, B10) and d (e.g., DBA/1, DBA/2) haplotypes. V alpha 11+ T cells are absent in strains with the a (e.g., BALB/c, C3H) or c (e.g., SJL) haplotypes of the Tcra gene complex. Finally, V alpha 11 is expressed predominantly on CD4+ T cells.

The RR8-1 antibody recognizes the V alpha 11.1 and 11.2 alleles, but not 11.3. This antibody has been reported to stimulate V alpha 11+ T cells.
Description & Specifications

Specifications

V alpha 11 TCR
Rat
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
RR8-1
Valpha11, Va11
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer