BUCHI V-100 Vacuum Pump
The Vacuum Pump V-100 is a chemically resistant PTFE diaphragm vacuum pump with space saving design. It covers essential needs in vacuum generation.
$4537.81 - $10308.55
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
05405207
|
BUCHI
11V100111 |
I-100 Interface, Woulff Bottle, and Secondary Condenser with Insulation |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
05-405-121
|
BUCHI
11V100000 |
No Optional Equipment |
Each for $4,537.81
|
|
|||||
05-405-118
|
BUCHI
11V100010 |
Woulff Bottle (No Interface or Secondary Condenser) |
Each for $4,866.14
|
|
|||||
05405115
|
BUCHI
11V100110 |
I-100 Interface and Woulff Bottle (No Secondary Condenser) |
Each for $7,879.82
|
|
|||||
Description
- Effective vacuum source for limited budgets
- Small footprint saves valuable laboratory space
- Light weight and compact design with integrated carrying handle
- High up time due to low maintenance
- Accurate vacuum regulation thanks to optional interface
- Maximum longevity due to: Robust and stable design and Integrated ECO2 mode
- On/off-regulated by the Interface I-100, optimally suited to work with Rotavapor R-100
- Includes silencer and 2m of vacuum tubing
Specifications
IP 20 | |
100/240 V |