Please login to your online account to display your discounted pricing
UVP Sample Plates and Plate Holder for GelMax Imager
Maximize imaging capabilities by adding interchangeable sample plates to view a wide range of fluorophore and colorimetric stains
Manufacturer: uvp  38038102
 View more versions of this product
Catalog No. UVP38038102
  • / Each
Description & Specifications
Specifications
Product Type Visi Blue Interchangeable Sample Plate
For Use With (Equipment) Gelmax imager
Wavelength 302nm UV to 460-470nm
For Use With (Application) Maximize imaging capabilities by adding interchangeable sample plates to view a wide range of fluorophore and colorimetric stains