missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

UVP Portable UV Lamp Replacement Tubes

Provide a variety of uses including laboratory, research, or criminology applications. UVP™ Portable UV Lamp Replacement Tubes are for use with Model ML-49 Portable UV Lamp with Handle.

Manufacturer:  UVP 34001501

 View more versions of this product

Catalog No. UVP34001501


 

Specifications

Description

Tube multiband shortwave / longwave accessory to handheld UV lamps 6W

Lamps are lightweight for easy handling
  • Durable plastic housing
  • Six watts
  • Models available with shortwave, longwave UV or a combination shortwave/longwave
  • Combination shortwave/longwave version comes with a snap-on wavelength selector for switching between wavelengths
Description & Specifications

Specifications

Tube multiband shortwave / longwave accessory to handheld UV lamps 6W
UV Lamp
6w
254/365nm; 6w
ML-49 Portable UV Lamp
Each
365nm Uv