Please login to your online account to display your discounted pricing
UVP MultiDoc-It™ Imaging Systems
Combines digital color camera, darkroom enclosure and transilluminator for digital imaging of gels, plates, and membranes in multiple-user laboratories
Manufacturer: uvp  97019201
 View more versions of this product
Computer for imaging and camera control. USB port required for camera connection; CD-ROM drive required for loading software; Windows™ 98SE, ME, XP or 200 operating system.
Catalog No. UVP97019201
  • / Each
Description & Specifications
Specifications
Product Type MultiDoc-It Imaging System
Transilluminator M-20
Wavelength 302nm
For Use With (Application) Fluorescent and colorimetric imaging
Voltage 115V