missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

UVP Mini UV Lamps

Battery operated and used as portable lamp for industrial, laboratory, lapidary and field inspections.

Supplier:  UVP 95012505DUP

Catalog No. 11-990-76



Description

Description

  • Battery-operated 4 watt lamps with lightweight, pocket-size design
  • Features impact-resistant plastic housing with attached nylon strap
  • Select from models with 254nm or 365nm UV or combination of both
  • Applications include fluorescence and inspection for field use
Specifications

Specifications

6.25 in.
Handheld mini UV lamp
Fluorescence and inspection for field use, industrial inspections, laboratory and lapidary
Each
1 in.
25mm
159mm
2.2 in.
Plastic
4w
6.25 x 1 in. (159 x 25mm)
56mm
365nm
SDS
Documents

Documents

Promotions

Promotions