UVP GelMax Imager
View, capture, analyze and document precast and mini-gel data
Specifications
Camera Type Color; 15.1 Megapixels
Filter Type 5-position tray, rotary knob controlled; EtBr filter included;additional filters available
Software Acquisition and analysis (compatible with Win 7, XP, Vista)
Transilluminator Built-in 302nm UV, 11.5 x 16cm plate size
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
UVP97067201 UVP
97067201
Imager W/O Free Visi-Blue or White Light plates N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
GelMax UVPCPQ8700
  • Fast, easy imaging of wide range of precast and mini gels up to 11.5 x 16cm
  • Includes darkroom with color camera, epi white light, 302nm UV transilluminator, black plate and Doc-ItLS acquisition, quantitative analysis software
  • White light Plate for viewing protein gels such as Coomassie Blue and Silver stains
  • Visi-Blue™ Plate emits 460-470nm blue light for viewing stains such as SYBR Green, SYBR Safe and GelGreen
  • Compact design reduces lab space requirements
  • Safety switch shut off of transillumination lighting after 10 minutes or with open drawer
GelMax UVP970672-0
  • Fast, easy imaging of wide range of precast and mini gels up to 11.5 x 16cm
  • Includes darkroom with color camera, epi white light, 302nm UV transilluminator, black plate and Doc-ItLS acquisition, quantitative analysis software
  • Compact design reduces lab space requirements
  • Safety switch shut off of transillumination lighting after 10 minutes or with open drawer
Order Info
  • Sample Visi-Blue™ and White Light, Converter Visi-Blue™ and White Light, Longwave 365nm plates and Plate Holders are also available separately
  • Computer/monitor not included but available separately
Description & Specifications
Specifications
Color; 15.1 Megapixels
5-position tray, rotary knob controlled; EtBr filter included;additional filters available
Acquisition and analysis (compatible with Win 7, XP, Vista)
Built-in 302nm UV, 11.5 x 16cm plate size