UVP GelMax Imager
View, capture, analyze and document precast and mini-gel data
Manufacturer:  UVP 97067201
 View more versions of this product
Catalog No. UVP97067201
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
Imager W/O Free Visi-Blue or White Light plates
Built-in 302nm UV, 11.5 x 16cm plate size
Color; 15.1 Megapixels
Acquisition and analysis (compatible with Win 7, XP, Vista)
15.1 MP
5-position tray, rotary knob controlled; EtBr filter included;additional filters available