Please login to your online account to display your discounted pricing
UVP GelDoc-It2 310 Imaging System
Captures high resolution images and facilitates quantitative analysis of fluorescent gels
Manufacturer: uvp  97027301
 View more versions of this product
Computer/monitor need to be ordered separately
Catalog No. UVP97027301
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Product Type GelDoc-It2 310 Imaging System
Transilluminator 254, 302, 365nm
Lens Type Manual lens: 12.5-75mm f/1.2
For Use With (Application) Image capturing and quantitative gels analysis
Camera Type 2 megapixel
Filter Type EtBr