UVP GelDoc-It™ DNA and Protein Gel Imaging System
Specifications
Compatibility Windows™ 7, XP, Vista
Depth (English) 14.5 in.
Depth (Metric) 36.8cm
Filter Type Ethidium Bromide Broadband
For Use With (Application) DNA and protein gel imaging, blue light gels, TLC gels, colony plates, visible light gels, autoradiograph: fluorescence, colorimetric
View More Specs
Products 1
Catalog Number Mfr. No. Wavelength Price Quantity  
Catalog Number Mfr. No. Wavelength Price Quantity  
UVP97057701 View Documents UVP
97057701
UV: 254/302/365nm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Windows™ 7, XP, Vista
36.8cm
DNA and protein gel imaging, blue light gels, TLC gels, colony plates, visible light gels, autoradiograph: fluorescence, colorimetric
33.5 in.
FluorCam 220 camera/lens kit, darkroom with gel viewing window, epi white light, Ethidium Bromide filter, choice of transilluminator and VisionWorksLS capture and analysis software
1280 x 1024
1.3MP, extendable to 5MP
17.5 in.
14.5 in.
Ethidium Bromide Broadband
0 to 65, 535
85.1cm
8 to 48mm for 1.2 manual zoom
GelDoc-Ite Imaging System
VisionWorksLS image acquisition and analysis, single-user license
44.4cm