Please login to your online account to display your discounted pricing
UVP Filter Assemblies for Handheld UV Lamps
Manufacturer: uvp  98000403
 View more versions of this product
Catalog No. UVP98000403
  • / Each
Description & Specifications
Specifications
Wavelength 254nm and 365nm
Product Type Filter assembly
For Use With (Equipment) Model UVGL-58 handheld UV lamp
Quantity Each