Please login to your online account to display your discounted pricing

UVP Compact and Handheld UV Lamps


Essential multipurpose UV lamps for laboratory use

Manufacturer: uvp  95002112

 View more versions of this product

Catalog No. UVP95002112

  • / Each
 

Description & Specifications

Specifications

Voltage 115V
Electrical Requirements 115V 60Hz
Hertz 60Hz
Lighting Intensity 760/720μw/cm2
Lighting Type Compact
Wattage 4w
Wavelength 254/365nm
Wavelength Range 365 to 254nm
Quantity Each
Case Material Plastic