Please login to your online account to display your discounted pricing

UVP Clear UV Transmitting Covers


Manufacturer: uvp  19013801

 View more versions of this product

Catalog No. UVP19013801

  • / Each
 

Description & Specifications

Specifications

Product Type Uv Transmitting Cover
Color Clear
For Use With (Equipment) For use with xx-20 series benchtop uv lamps