Please login to your online account to display your discounted pricing

UVP Clear UV Blocking Covers


Manufacturer: uvp  19012101

 View more versions of this product

Catalog No. UVP19012101

  • / Each
 

Description & Specifications

Specifications

Product Type Uv Blocking Cover
For Use With (Equipment) 3uv™ transilluminators
Color Clear