UVP ChemiDoc-It2™ 810 Imager
For high resolution, high sensitivity image capture and extensive quantitative analysis of chemiluminescent blots and fluorescent gels
Manufacturer:  UVP 97065101
 View more versions of this product
Catalog No. UVP97065101
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
CSA
15 in.
50mm
81cm
UVP Imager with M-26XV Transilluminator
115V
18.5 in.
VisionWorks LS
Windows™ XP (SP2), Win 7 (32-bit)
38cm
32 in.
MegaCam810 cooled CCD camera/lens, EtBr filter, UV transilluminator, VisionWorksLS Acquisition and Analysis Software
8.1 MXP
302nm
45cm
Documents