Please login to your online account to display your discounted pricing
UVP BioSpectrum™ 510 Imaging Systems
Advanced, automated image capture of gels and blots for multiple applications
Catalog Number Mfr. No. Lens Type Wavelength Filter Type Price Quantity    
12.5-75mm f/1.2 254/302/365nm 21 x 26cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to for assistance.
12.5-75mm f/1.2 302nm 25 x 26cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to for assistance.
Description & Specifications
  • Comes with high resolution scientific grade deeply cooled CCD camera
  • VisionWorks™LS software for darkroom control, image capture, enhancement and analysis
  • Light tight dark room with gel viewing window and wide access door for visualization of samples
  • Built-in epi lighting (365nm UV, 460-470nm blue light and broadband white light) enables overhead excitation and illumination of samples
  • Choice of UV transilluminator models for ultraviolet excitation plus LED white light plate for viewing colonies, Coomassie Blue and Silver stained gels
  • No film required for chemiluminescent western blot imaging
  • Manually controlled platform lift
  • Emission filters: Five position motorized wheel with EtBr (580 to 630nm), SYBR Green (510 to 560nm), SYBR Gold (520 to 620nm)
  • Chemi tray fits on transilluminator for placement of chemiluminescent blots
Camera 510
  • Regulated cooling and low noise for imaging variety of chemiluminescent and fluorescent samples
  • 16 bit, 4.1 megapixel resolution extendable to 12MP
  • Cooling at -35°C from ambient
  • Lens: 12.5 to 75mm f/1.2, 25mm f/0.95, motorized zoom
Software licenses for multi-user and networks are available. PC/monitor ordered separately.
  • For chemiluminescence, fluorescence and colorimetric as well as NIR and multiplex applications
  • In-depth automated analysis allows generation of 1D quantitation, lane profile graphs, histograms, area density, western blot densitometry, colony counting, multiplex etc.