Please login to your online account to display your discounted pricing

UVI 3003, Tocris

RXR antagonist

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material UVI 3003
Assay Percent Range >98%
Molecular Formula C28H36O4
Formula Weight 436.58
Synonym 3-[4-Hydroxy-3-[5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-(pentyloxy)-2-naphthalenyl]phenyl]-2-propenoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

330310

 
r & d systems
RDS330310
10mg Each for $214.12

330350

 
r & d systems
RDS330350
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 436.58
Quantity 10mg
Synonym 3-[4-Hydroxy-3-[5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-(pentyloxy)-2-naphthalenyl]phenyl]-2-propenoic acid
Molecular Formula C28H36O4
CAS 847239-17-2
Chemical Name or Material UVI 3003
Assay Percent Range >98%
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C