Honeywell™ Uvex™ XC™ Safety Glasses Replacement Lenses
Extended wrap-around lens for extra cheek and extra side protection. Soft flexible nose fingers conform to fit a variety of nasal profiles. Adjustable temples and lens inclination.
Manufacturer: Honeywell™ UVS6951
Description
Uvex Genesis XC has the same comfort and adjustability features as original Genesis, with 20% more extended wrap-around lens for extra cheek and side coverage. Rx lens insert available. Uvex Genesis XC has an easy and economical lens replacement system.
- PC lens, Hydroshield AF, Dura-streme AF/HC
- Available in a very wide range of lens tints
- 20% more coverage than original Genesis
- Economical lens replacement system
- Meets military Vo ballistic impact requirement (on select tints)
Specifications
XC Spectacles | |
Ultradura | |
Gray | |
Replacement lens |