Honeywell™ Uvex™ XC™ Safety Glasses Replacement Lenses
Extended wrap-around lens for extra cheek and extra side protection. Soft flexible nose fingers conform to fit a variety of nasal profiles. Adjustable temples and lens inclination.
Manufacturer:  Honeywell™ UVS6950X
 View more versions of this product
Catalog No. 19-066-044
Description
Description
Uvex Genesis XC has the same comfort and adjustability features as original Genesis, with 20% more extended wrap-around lens for extra cheek and side coverage. Rx lens insert available. Uvex Genesis XC has an easy and economical lens replacement system.
  • PC lens, Hydroshield AF, Dura-streme AF/HC
  • Available in a very wide range of lens tints
  • 20% more coverage than original Genesis
  • Economical lens replacement system
  • Meets military Vo ballistic impact requirement (on select tints)
Specifications
Specifications
XC Spectacles
Uvextreme
Clear
Replacement lens