Honeywell™ Uvex™ VersaPro™ Eyewear
Extended-wear comfort features and increased durability
$133.01 - $133.01
Specifications
Certifications/Compliance Meets ANSI Z87.1-2003 (High Impact) standard. Certified to the requirements of CSA Z94.3 standard.
Frame Color Blue, Black
Frame Material Nylon
Includes Flexible and replaceable nosepiece
Temple Style Ratcheting, FlexFit bendable
View More Specs
Products 1
Catalog Number Mfr. No. For Use With (Application) Lens Coating Lens Tint Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Lens Coating Lens Tint Price Quantity  
18-813-447
View Documents
Honeywell™
SX0102
Ideal for low-light applications where enhanced contrast is needed Supra-Dura Amber
Pack of 10 Each for $133.01
Description
Description
  • Sleek, streamlined lenses provide excellent coverage and protection
  • FlexFit wire core temples bend and flex for a customizable, full-comfort fit
  • Flexible and replaceable nosepiece conforms to virtually any nasal profile and provides a contoured, non-slip fit
  • Ratcheting temples enable wearers to adjust lens inclination angle to fit perfectly above cheekbones
Compliance
Exceeds ANSI™ Z87.1-2003.
Specifications
Specifications
Meets ANSI Z87.1-2003 (High Impact) standard. Certified to the requirements of CSA Z94.3 standard.
Nylon
Ratcheting, FlexFit bendable
Blue, Black
Flexible and replaceable nosepiece
SDS