Uvex™ Stealth™ OTG Goggles: Replacement Parts and Accessories
Replacement lenses and accessories for Uvex Stealth OTG goggles
$7.74 - $7.74
Specifications
Product Type Replacement Lens
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Lens Coating Lens Tint Price Quantity  
Catalog Number Mfr. No. Lens Coating Lens Tint Price Quantity  
18-813-429 North Safety Products
S740D
dura-streme anti-fog/anti-scratch Clear Each for $7.74
Pack of 10 Each for $75.10
Specifications
Description
For use with Uvex Stealth OTG goggles (Cat. No. 18-813-533)
  • Ideal for impact, dust/airborne particles, chemical splash, and optical radiation (UV and welding/heat radiation)
  • Superior optics and panoramic field of vision enhanced with toric lens
  • Pivoting clip and quick-adjusting wide headband
  • Elastomer body conforms and contours for a secure fit—even over most prescription eyewear
  • Replacement lenses easily snap on
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards.
Description & Specifications
Specifications
Replacement Lens