Honeywell Safety Products™ Uvex™ Skyper™ Safety Glasses
Custom fit with superior peripheral protection
Manufacturer:  Honeywell Safety Products™ S1908
 View more versions of this product
Catalog No. 29-636-1686
Description
Description
Custom fit with superior peripheral protection
  • Polycarbonate lenses with single-lens design feature Ultradura™ hardcoat (UD) or Uvextreme™ AF (XTR) antifog, antiscratch, and antistatic coating
  • Wraparound, impact resistant lenses block 99.9% of harmful UVA and UVB radiation
  • Black frames with molded-in browguard reduce the danger of flying particulates
  • Molded-in nosebridge offers comfort and safety in a variety of work applications
  • Patented, adjustable, comfort-cushioned Duoflex™ temples
  • Simple lens replacement system, ratcheting lens inclination
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3-1999 standards.
Specifications
Specifications
Meet ANSI Z87.1-2003 and CSA Z94.3-1999 standards.
Black
Impact-resistant,IR Shade 5,Scratch-resistant
Shade 5.0 Infra-dura
Shade 5.0