Honeywell™ Uvex™ Seismic™ Sealed Eyewear
Eye protection against impact, sun, wind, dust and airborne debris
Supplier:  Honeywell™ S0609HS
 View more versions of this product
Catalog No. 19-185-726
Description
Description
  • For dirty, high-particulate work environments
  • With Dura-streme dual coating technology which combines two types of lens coatings - Uvextreme anti-fog (interior) and Ultra-dura anti-scratch (exterior), as well as Uvextra AF anti-fog/Hardcoat anti-scratch coating options
  • Soft and flexible nasal pads
  • Soft, high-quality face cushioning and padded temple tips
  • Highly adjustable flame-resistant headband
  • Replaceable flame-resistant, cushion-lined subframe
Applications:
For applications like grinding, chipping, shaving and sanding.
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Specifications
SCT LOW IR
For Applications Like Grinding, Chipping, Shaving and Sanding
Black
Removable