Honeywell™ Uvex™ Protective Spectacle Cases
Manufacturer: Honeywell™ UVS491
Description
Attractive cases protect safety spectacles from scratches and breakage when not in use. Textured, easy-to-clean cases come in five styles: oversized black vinyl case holds larger spectacles; sport style nylon case holds larger spectacles and comes with drawstring and barrel lock; hard case offers extra protection for metal frame eyewear, with belt loop and Velcro™ closure; roomy nylon goggle pack accommodates all Uvex goggles, with two belt loops and zippered closure; and similarly styled nylon Astro Pack is designed to hold Astrospec™ eyewear.
Specifications
Eyewear Pack | |
Astrospec™ eyewear | |
Eyewear case |
Protect safety spectacles from scratches and breakage when not in use | |
Nylon |