Honeywell™ Uvex™ Genesis XC™ Protective Eyewear
Extended wraparound lens provides excellent coverage
Manufacturer:  Honeywell™ UVS3307
 View more versions of this product
Catalog No. 19-066-040
Description
Description
  • Soft, flexible nose fingers conform to fit a variety of nasal profiles
  • Black frames with adjustable temples for comfortable fit; cushioned for all-day comfort
  • Available lens tints include clear, gray, mirror, Infradura™ welding shades, and SCT™ to reduce spectral hazards
  • All clear lenses absorb >99.9% of UVA and UVB radiation up to 385nm; tinted lenses absorb >99.9% UVA and UVB radiation up to 400nm
  • Rx Insert available (see cat. no. 191300825)
uvextreme™ Anti-Fog Coating (XTR)
  • Permanently bonded to lens
  • Absorbs 99.9% of UV radiation
  • Anti-scratch and -static
  • Will not wear after repeated cleanings
ultra-dura™ Hard Coat (UD)
  • Permanently bonded to lens
  • Scratch- and chemical-resistant
dura-streme™ Combination Coating (DS)
  • ultra-dura coated exterior; uvextreme coated interior
  • Lengths lens life
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards.
1Reduces glare for workers moving between indoor to outdoor work environments.
2Provide broad protection against accidental arcs. Equivalent to a Shade 2.0. welding filter, but not designed for any welding applications.
3Provide protection from short-term electric arcs and radiation generated when an arc accidentally occurs. Equivalent to a Shade 1.2 welding lens, but not designed for any welding applications.
Specifications
Specifications
Meet ANSI Z87.1-2003 and CSA Z94.3 standards.
Black
Shade 5.0 Infradura
Shade 5.0 Infra-dura
Cushioned
IR Shade 5,Scratch-resistant