Honeywell™ Uvex™ Genesis™ Safety Glasses, Replacement Lenses
Wrap-around lens provides exceptional peripheral vision and protection. Flexible fingers adjust to most nasal profiles. Elastomer browguard deflects impact. Adjustable temples and lens inclination.
Manufacturer: Honeywell™ UVS6901X
Description
Uvex Genesis is engineered to deliver on the best all-day comfort, protection and style. It's built with MMT for comfort, and protects in a very wide assortment of lens tints. Uvex Genesis has an easy and economical lens replacement system. Frames have a lifetime guarantee. Made in the USA.
- PC lens, Hydroshield AF, Dura-streme AF/HC, Uvextreme
- Ratchet, adjustable temples, flexible nose, MMT
- Available in a very wide range of lens tints
- Economical lens replacement system
Specifications
Safety Spectacles | |
Uvextreme Anti-fog | |
Replacement lens |
genesis Spectacles | |
Espresso |