Honeywell™ Uvex™ Genesis™ Safety Glasses, Replacement Lenses
Wrap-around lens provides exceptional peripheral vision and protection. Flexible fingers adjust to most nasal profiles. Elastomer browguard deflects impact. Adjustable temples and lens inclination.
Manufacturer:  Honeywell™ UVS6902X
 View more versions of this product
Catalog No. 29-635-2405
Description
Description
Uvex Genesis is engineered to deliver on the best all-day comfort, protection and style. It's built with MMT for comfort, and protects in a very wide assortment of lens tints. Uvex Genesis has an easy and economical lens replacement system. Frames have a lifetime guarantee. Made in the USA.
  • PC lens, Hydroshield AF, Dura-streme AF/HC, Uvextreme
  • Ratchet, adjustable temples, flexible nose, MMT
  • Available in a very wide range of lens tints
  • Economical lens replacement system
Specifications
Specifications
Replacement Lenses/Spectacles
Uvextreme Anti-fog
Replacement lens
genesis Spectacles
Amber