Honeywell™ Uvex™ Genesis™ Reading Magnifier Safety Glasses
High-impact protection
Supplier: Honeywell™ S3761
Description
- Soft, flexible nasal fingers
- Padded temple tips and cushioned browbar
- Adjustable temple length and ratcheting lens inclination provides for secure fit
- Easy and economical lens replacement (see Cat. No. 18-813-424)
- With Ultra-dura™ antiscratch coating
- Five diopter strengths available
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards.
Specifications
Ultra-dura Scratch-resistant | |
Clear | |
Ideal for most indoor work applications | |
+1.50 |
ANSI Z87.1-2015, CSA Z94.3-2007 | |
Reading Magnifier Spectacles | |
Black | |
0.999 |