Honeywell™ Uvex™ Genesis™ Protective Safety Glasses
Three-position ratchet lens inclination system and adjustable temples enable wearer to customize fit
$11.56 - $170.42
Catalog Number | Mfr. No. | Frame Color | Lens Tint | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Frame Color | Lens Tint | Price | Quantity | ||||
19-003-283
|
Honeywell™
UVS3207 |
Black | IR Filter Shade 3.0 |
Pack of 10 Pair for $170.42
|
|
||||
19-040-143
|
Honeywell™
UVS3200 |
Black | Clear |
Each for $13.77
Pack of 10 Each for $133.60
|
|
||||
19-040-145
|
Honeywell™
UVS3201 |
Black | Espresso |
Each for $16.05
Pack of 10 Each for $155.70
|
|
||||
19-040-152
|
Honeywell™
UVS3204 |
Black | SCT-Reflect 50 |
Each for $18.88
Pack of 10 Each for $183.10
|
|
||||
19-040-156
|
Honeywell™
UVS3221 |
Earth | Espresso |
Pack of 10 Each for $95.03
|
|
||||
19-040-158
|
Honeywell™
UVS3222 |
Earth | Amber |
Pack of 10 Each for $92.12
|
|
||||
19-040-157
|
Honeywell™
UVS3221X |
Earth | Espresso |
Each for $11.56
Pack of 10 Each for $112.10
|
|
||||
19-040-170
|
Honeywell™
UVS3241 |
Vapor Blue | Espresso |
Each for $14.26
Pack of 10 Each for $138.30
|
|
||||
19-040-171
|
Honeywell™
UVS3241X |
Vapor Blue | Espresso |
Each for $22.09
Pack of 10 Each for $214.20
|
|
||||
19-040-168
|
Honeywell™
UVS3240 |
Vapor Blue | Clear |
Each for $13.77
Pack of 10 Each for $133.60
|
|
||||
19-040-172
|
Honeywell™
UVS3242 |
Vapor Blue | Amber |
Each for $12.38
Pack of 10 Each for $120.10
|
|
||||
19-040-173
|
Honeywell™
UVS3243 |
Vapor Blue | Mirror |
Each for $18.26
Pack of 10 Each for $177.00
|
|
||||
19-820-987
|
Honeywell™
UVS3202X |
Black | Amber |
Each for $22.09
Case of 10 Each for $214.20
|
|
||||
19-185-703
|
Honeywell™
S3240HS |
Vapor Blue | Clear |
Each for $15.37
Case of 200 Each for $2,810.00
|
|
||||
Description
- Impact-resistant polycarbonate lenses block >99.9% of harmful UVA and UVB radiation
- Ventilation channel between frame and lens prevents moisture buildup
- Nose bridge is covered with small, ultrasoft “fingers” that hug the nose for improved fit and comfort
- Soft elastomer browguard cushions all points of contact with face
uvextreme™ Anti-Fog Coating (XTR)
- Permanently bonded to lens
- Absorbs 99.9% of UV radiation
- Anti-scratch and -static
- Will not wear after repeated cleanings
- Provides 99.9% ultra-violet protection
- Provides 2X greater scratch-resistance
- Extended wrap-around lens for extra cheek and extra side protection
- Soft flexible nose fingers conform to fit a variety of nasal profiles
- Adjustable temples and lens inclination
- Permanently bonded to lens
- Scratch- and chemical-resistant
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards. Meet stringent military V0 Ballistic Test for Impact requirements.
Warranty and Services
Lifetime frame guarantee.
Specifications
Lifetime frame guarantee |