Fisher Scientific Uvex™ Futura™ Goggles
180° panoramic view for better peripheral vision
Supplier: Fisher Scientific S47619C
Description
- Firm, comfortable seal protects against chemicals, dust, and grindings
- Impact-resistant, chemical-splash polycarbonate lens
- Permanently bonded uvextreme™ AF antifog coating
- Wide, neoprene strap is slotted to prevent slippage
Compliance
Meet ANSI Z87.1-2010 and CSA Z94.3 standards
Specifications
ANSI Z87.1-2010 and CSA Z94.3 standards | |
Clear | |
Uvextreme Anti-fog |
Anti-fog | |
Safety goggles |